accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OFHNONHOO

CAS RN 105637-38-5

IUPAC Name: benzyl N-{1-[(4-fluoro-3-oxobutan-2-yl)carbamoyl]-2-phenylethyl}carbamate
Synonyms: Z-FA-fmk
Molecular Weight (g/mol): 386.42
Molecular Formula: C21H23FN2O4
InChi Key: ASXVEBPEZMSPHB-UHFFFAOYNA-N
SMILES: CC(NC(=O)C(CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1)C(=O)CF
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results