accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 105637-50-1

HClClSOONHN

CAS RN 105637-50-1

IUPAC Name: hydrogen 1-[(5-chloronaphthalen-1-yl)sulfonyl]-1,4-diazepane chloride
Synonyms: hydrogen 1-(5-chloronaphthalen-1-ylsulfonyl)-1,4-diazepane chloride
Molecular Weight (g/mol): 361.28
Molecular Formula: C15H18Cl2N2O2S
InChi Key: ZNRYCIVTNLZOGI-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=C2C=CC=C(C2=CC=C1)S(=O)(=O)N1CCCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results