accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOH

CAS RN 105640-07-1

IUPAC Name: 4-(4-hydroxyphenyl)cyclohexan-1-one
Synonyms: 4-(4-hydroxyphenyl)cyclohexan-1-one
Molecular Weight (g/mol): 190.24
Molecular Formula: C12H14O2
InChi Key: SLJYPZJZQIHNGU-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1CCC(=O)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results