accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2O(R)(S)H3N(R)(S)

CAS RN 105786-40-1

IUPAC Name: (1R,2S,3R,4S)-3-carbamoylbicyclo[2.2.1]hept-5-en-2-aminium
Synonyms: (1R,2S,3R,4S)-3-carbamoylbicyclo[2.2.1]hept-5-en-2-aminium
Molecular Weight (g/mol): 153.2
Molecular Formula: C8H13N2O
InChi Key: SCQSHSJVMGGQKR-UCROKIRRSA-O
SMILES: NC(=O)[C@H]1[C@@H]([NH3+])[C@@H]2C[C@H]1C=C2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)