accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10581-38-1

IUPAC Name: 2,2,6,6-tetramethylpiperidin-4-one hydrate
Synonyms: triacetone amine hydrate
Molecular Weight (g/mol): 173.26
Molecular Formula: C9H19NO2
InChi Key: SFDODBGIDPNFEH-UHFFFAOYSA-N
SMILES: O.CC1(C)CC(=O)CC(C)(C)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results