accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10595-95-6

CH3NH3CNO

CAS RN 10595-95-6

IUPAC Name: ethyl(methyl)nitrosoamine
Synonyms: N-methyl-N-nitrosoethamine
Molecular Weight (g/mol): 88.11
Molecular Formula: C3H8N2O
InChi Key: RTDCJKARQCRONF-UHFFFAOYSA-N
SMILES: CCN(C)N=O
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results