accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHNOHOH(E/Z)

CAS RN 105955-11-1

IUPAC Name: N-[2-(3,4-dihydroxyphenyl)ethyl]octadec-9-enamide
Synonyms: N-[2-(3,4-dihydroxyphenyl)ethyl]octadec-9-enamide
Molecular Weight (g/mol): 417.63
Molecular Formula: C26H43NO3
InChi Key: QQBPLXNESPTPNU-UHFFFAOYSA-N
SMILES: CCCCCCCCC=CCCCCCCCC(=O)NCCC1=CC(O)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results