accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3

CAS RN 106-18-3

IUPAC Name: butyl dodecanoate
Synonyms: dodecanoic acid, butyl ester
Molecular Weight (g/mol): 256.43
Molecular Formula: C16H32O2
InChi Key: NDKYEUQMPZIGFN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results