accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3CH3OH

CAS RN 106-21-8

IUPAC Name: 3,7-dimethyloctan-1-ol
Synonyms: dihydrocitronellol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: PRNCMAKCNVRZFX-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)CCO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)