accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3CH3

CAS RN 106-27-4

IUPAC Name: 3-methylbutyl butanoate
Synonyms: isoamyl butyrate
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: PQLMXFQTAMDXIZ-UHFFFAOYSA-N
SMILES: CCCC(=O)OCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results