accessibility menu, dialog, popup

Test

CAS RN 106-30-9

CAS RN 106-30-9

IUPAC Name: ethyl heptanoate
Synonyms: wine oil aether oenanthicus ethyl heptanoate (natural) enanthylic ether ethyl oenanthylate ethyl hepanoate grape oil cognac oil oenanthic ether oleum vitis viniferae
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: TVQGDYNRXLTQAP-UHFFFAOYSA-N
SMILES: CCCCCCC(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results