accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3

CAS RN 106-36-5

IUPAC Name: propyl propanoate
Synonyms: propyl propanoate N-propylpropionate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: MCSINKKTEDDPNK-UHFFFAOYSA-N
SMILES: CCCOC(=O)CC
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results