accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHBr

CAS RN 106-41-2

IUPAC Name: 4-bromophenol
Synonyms: 4-bromophenol P-bromophenol ccris 632 C6H5BrO
Molecular Weight (g/mol): 173.01
Molecular Formula: C6H5BrO
InChi Key: GZFGOTFRPZRKDS-UHFFFAOYSA-N
SMILES: OC1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)