accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3Cl

CAS RN 106-43-4

IUPAC Name: 1-chloro-4-methylbenzene
Synonyms: P-chlorotoluene C7H7cl 4-chlorotoluene, ion(1+) 38cl-labeled 4-chlorotoluene ion(1+) 4-chlorotoluene 4-chlorotoluene (substance) 4-chlorotoluene, 38cl-labeled
Molecular Weight (g/mol): 126.58
Molecular Formula: C7H7Cl
InChi Key: NPDACUSDTOMAMK-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)