accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2Cl

CAS RN 106-47-8

IUPAC Name: 4-chloroaniline
Synonyms: 4-chloroaniline caswell no. 182 rcra waste number P024 epa pesticide chemical code 017203 ccris 131 C6H6ClN p-CA
Molecular Weight (g/mol): 127.57
Molecular Formula: C6H6ClN
InChi Key: QSNSCYSYFYORTR-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (11)
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (8)
Concentration 
  • (3)
  • (3)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (1)
  • (11)
  • (9)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (8)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)