accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SHCl

CAS RN 106-54-7

IUPAC Name: 4-chlorobenzene-1-thiol
Synonyms: 4-chlorobenzenethiol
Molecular Weight (g/mol): 144.62
Molecular Formula: C6H5ClS
InChi Key: VZXOZSQDJJNBRC-UHFFFAOYSA-N
SMILES: SC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (4)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)