accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 106-63-8

IUPAC Name: 2-methylpropyl prop-2-enoate
Synonyms: isobutyl acrylate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: CFVWNXQPGQOHRJ-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results