accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOCH3

CAS RN 106-65-0

IUPAC Name: 1,4-dimethyl butanedioate
Synonyms: dimethyl succinate
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: MUXOBHXGJLMRAB-UHFFFAOYSA-N
SMILES: COC(=O)CCC(=O)OC
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)