accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 106-88-7

H3CO

CAS RN 106-88-7

IUPAC Name: 2-ethyloxirane
Synonyms: 1,2-butyleneoxide
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: RBACIKXCRWGCBB-UHFFFAOYNA-N
SMILES: CCC1CO1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)