accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 106243-16-7

SHNNNHN

CAS RN 106243-16-7

IUPAC Name: N-cyclohexyl-4-(1H-imidazol-5-yl)piperidine-1-carbothioamide
Synonyms: thioperamide
Molecular Weight (g/mol): 292.45
Molecular Formula: C15H24N4S
InChi Key: QKDDJDBFONZGBW-UHFFFAOYSA-N
SMILES: S=C(NC1CCCCC1)N1CCC(CC1)C1=CN=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results