accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOHOHClHCl

CAS RN 106261-49-8

IUPAC Name: 4-[(4-methylpiperazin-1-yl)methyl]benzoic acid dihydrochloride
Synonyms: 4-[(4-methylpiperazin-1-yl)methyl]benzoic acid dihydrochloride
Molecular Weight (g/mol): 307.22
Molecular Formula: C13H20Cl2N2O2
InChi Key: ISHROKOWRJDOSN-UHFFFAOYSA-N
SMILES: Cl.Cl.CN1CCN(CC2=CC=C(C=C2)C(O)=O)CC1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results