accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 106308-44-5

IUPAC Name: 1-[(2,6-difluorophenyl)methyl]-1H-1,2,3-triazole-4-carboxamide
Synonyms: rufinamide
Molecular Weight (g/mol): 238.2
Molecular Formula: C10H8F2N4O
InChi Key: POGQSBRIGCQNEG-UHFFFAOYSA-N
SMILES: NC(=O)C1=CN(CC2=C(F)C=CC=C2F)N=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results