accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 106391-86-0

CH3(R)HONHOOCH3CH3H3C

CAS RN 106391-86-0

IUPAC Name: tert-butyl N-(1-hydroxypropan-2-yl)carbamate
Synonyms: tert-butyl N-(1-hydroxypropan-2-yl)carbamate
Molecular Weight (g/mol): 175.23
Molecular Formula: C8H17NO3
InChi Key: PDAFIZPRSXHMCO-UHFFFAOYNA-N
SMILES: CC(CO)NC(=O)OC(C)(C)C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)