accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COFOCl

CAS RN 106428-06-2

IUPAC Name: 3-fluoro-2-methoxybenzoyl chloride
Synonyms: 3-fluoro-2-methoxybenzoyl chloride
Molecular Weight (g/mol): 188.58
Molecular Formula: C8H6ClFO2
InChi Key: WJLYPIGIRQBXFF-UHFFFAOYSA-N
SMILES: COC1=C(C=CC=C1F)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results