accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiCH3CH3HC

CAS RN 1066-54-2

IUPAC Name: ethynyltrimethylsilane
Synonyms: trimethylsilylacetylene
Molecular Weight (g/mol): 98.22
Molecular Formula: C5H10Si
InChi Key: CWMFRHBXRUITQE-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#C
Quantity 
  • (1)
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results