accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaOHNOOSOOO

CAS RN 106627-54-7

IUPAC Name: sodium 1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate
Synonyms: sodium 1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate
Molecular Weight (g/mol): 217.13
Molecular Formula: C4H4NNaO6S
InChi Key: RPENMORRBUTCPR-UHFFFAOYNA-M
SMILES: [Na+].ON1C(=O)CC(C1=O)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)