accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 106649-02-9

IUPAC Name: 1-methyl-1H-indazole-3-carbonyl chloride
Synonyms: 1-methylindazole-3-carbonyl chloride
Molecular Weight (g/mol): 194.62
Molecular Formula: C9H7ClN2O
InChi Key: NPNWVMBPSINFBV-UHFFFAOYSA-N
SMILES: CN1N=C(C(Cl)=O)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results