accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1067-25-0

CH3SiOH3COH3COH3C

CAS RN 1067-25-0

IUPAC Name: trimethoxy(propyl)silane
Synonyms: trimethoxy(propyl)silane
Molecular Weight (g/mol): 164.28
Molecular Formula: C6H16O3Si
InChi Key: HQYALQRYBUJWDH-UHFFFAOYSA-N
SMILES: CCC[Si](OC)(OC)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results