accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1068-22-0

NH4CH3OPSSOH3C

CAS RN 1068-22-0

IUPAC Name: ammonium O,O-diethyl sulfanidylphosphonothioate
Synonyms: ammonium diethyl dithiophosphate
Molecular Weight (g/mol): 203.25
Molecular Formula: C4H14NO2PS2
InChi Key: HFRHTRKMBOQLLL-UHFFFAOYSA-N
SMILES: [NH4+].CCOP([S-])(=S)OCC
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)