accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1068-55-9

CH3CH3MgCl

CAS RN 1068-55-9

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 102.84
Molecular Formula: C3H7ClMg
InChi Key: IUYHWZFSGMZEOG-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].C[CH-]C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results