accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONH2FFF

CAS RN 106877-21-8

IUPAC Name: 4-phenoxy-2-(trifluoromethyl)aniline
Synonyms: 4-phenoxy-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 253.22
Molecular Formula: C13H10F3NO
InChi Key: CWKQMUBTZIPBLG-UHFFFAOYSA-N
SMILES: NC1=C(C=C(OC2=CC=CC=C2)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results