accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 106896-49-5

CH3OONH2Br

CAS RN 106896-49-5

IUPAC Name: methyl 4-amino-3-bromobenzoate
Synonyms: methyl 4-amino-3-bromobenzoate
Molecular Weight (g/mol): 230.06
Molecular Formula: C8H8BrNO2
InChi Key: AIUWAOALZYWQBX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(N)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results