accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOPOHOOH2NOOCH3

CAS RN 1069-79-0

IUPAC Name: (2-aminoethoxy)[2,3-bis(octadecanoyloxy)propoxy]phosphinic acid
Synonyms: DSPE
Molecular Weight (g/mol): 748.08
Molecular Formula: C41H82NO8P
InChi Key: LVNGJLRDBYCPGB-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC(COP(O)(=O)OCCN)OC(=O)CCCCCCCCCCCCCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results