accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 107-39-1

CH3H2CCH3CH3H3C

CAS RN 107-39-1

IUPAC Name: 2,4,4-trimethylpent-1-ene
Synonyms: 2,4,4-trimethyl-1-pentene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: FXNDIJDIPNCZQJ-UHFFFAOYSA-N
SMILES: CC(=C)CC(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Boiling Point 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)