accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 107-59-5

IUPAC Name: tert-butyl 2-chloroacetate
Synonyms: tert-butyl 2-chloroacetate
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: KUYMVWXKHQSIAS-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CCl
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results