accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 107-66-4

IUPAC Name: dibutoxyphosphinic acid
Synonyms: di-N-butyl phosphate
Molecular Weight (g/mol): 210.21
Molecular Formula: C8H19O4P
InChi Key: JYFHYPJRHGVZDY-UHFFFAOYSA-N
SMILES: CCCCOP(O)(=O)OCCCC
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)