accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3Br

CAS RN 107-81-3

IUPAC Name: 2-bromopentane
Synonyms: 2-bromopentane
Molecular Weight (g/mol): 151.05
Molecular Formula: C5H11Br
InChi Key: LGAJYTCRJPCZRJ-UHFFFAOYNA-N
SMILES: CCCC(C)Br
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (2)