accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1070-11-7

ClClH3C(S)HONHHN(S)CH3OHHH

CAS RN 1070-11-7

IUPAC Name: dihydrogen (2S)-2-[(2-{[(2S)-1-hydroxybutan-2-yl]amino}ethyl)amino]butan-1-ol dichloride
Synonyms: dihydrogen ethambutol dichloride
Molecular Weight (g/mol): 277.23
Molecular Formula: C10H26Cl2N2O2
InChi Key: AUAHHJJRFHRVPV-BZDVOYDHSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CC[C@@H](CO)NCCN[C@@H](CC)CO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)