accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 107099-99-0

IUPAC Name: (2,5-dimethoxyphenyl)boronic acid
Synonyms: 2,5-dimethoxyphenylboronic acid
Molecular Weight (g/mol): 181.98
Molecular Formula: C8H11BO4
InChi Key: QOZLFNQLIKOGDR-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(=C1)B(O)O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)