CAS RN 107220-26-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac 3-Hydroxy-3-mercaptomethylquinuclidine, TRC
CAS: 107220-26-8 Molecular Formula: C8 H15 N O S Molecular Weight (g/mol): 173.28 Synonym: 3-(Mercaptomethyl)-1-azabicyclo[2.2.2]octan-3-ol,Cevimeline Impurity IUPAC Name: 3-(sulfanylmethyl)-1-azabicyclo[2.2.2]octan-3-ol SMILES: OC1(CS)CN2CCC1CC2