CAS RN 107220-28-0
CAS RN 107220-28-0
IUPAC Name:
hydrogen (2R,5'R)-5'-methyl-4-azaspiro[bicyclo[2.2.2]octane-2,2'-[1,4]oxathiolane] chloride
Synonyms:
hydrogen cevimeline chloride
Molecular Weight (g/mol):
235.77
Molecular Formula:
C10H18ClNOS
InChi Key:
SURWTGAXEIEOGY-GHXDPTCOSA-N
SMILES:
[H+].[Cl-].C[C@@H]1O[C@@]2(CS1)CN1CCC2CC1
1
–
4
of
4
results
1
Tocris Bioscience™ Cevimeline hydrochloride
CAS: 107220-28-0 Molecular Formula: C10H18ClNOS
2
rac,cis-Cevimeline Hydrochloride Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
rac,cis-Cevimeline Hydrochloride Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
rac,cis-Cevimeline Hydrochloride Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.