CAS RN 1072833-77-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ixazomib, TRC
CAS: 1072833-77-2 Molecular Formula: C14 H19 B Cl2 N2 O4 Molecular Weight (g/mol): 361.03 Synonym: Boronic acid, B-[(1R)-1-[[2-[(2,5-dichlorobenzoyl)amino]acetyl]amino]-3-methylbutyl]-,B-[(1R)-1-[[2-[(2,5-Dichlorobenzoyl)amino]acetyl]amino]-3-methylbutyl]boronic acid,Ixazomib,MLN 2238,Ninlaro IUPAC Name: [(1R)-1-[[2-[(2,5-dichlorobenzoyl)amino]acetyl]amino]-3-methylbutyl]boronic acid SMILES: CC(C)C[C@H](NC(=O)CNC(=O)c1cc(Cl)ccc1Cl)B(O)O