CAS RN 1072944-21-8
IUPAC Name:
[2-(methylsulfanyl)pyridin-3-yl]boronic acid
Synonyms:
2-(methylsulfanyl)pyridin-3-ylboronic acid
Molecular Weight (g/mol):
169.01
Molecular Formula:
C6H8BNO2S
InChi Key:
QLZLLRKAZYHWTE-UHFFFAOYSA-N
SMILES:
CSC1=C(C=CC=N1)B(O)O
1
–
1
of
1
results
1
2-(Methylthio)pyridine-3-boronic acid, 95%, Thermo Scientific™
CAS: 1072944-21-8 Molecular Formula: C6H8BNO2S Molecular Weight (g/mol): 169.005 MDL Number: MFCD09037488 InChI Key: QLZLLRKAZYHWTE-UHFFFAOYSA-N Synonym: 2-methylthio pyridine-3-boronic acid,2-methylthio pyridin-3-yl boronic acid,2-methylsulfanyl pyridin-3-yl boronic acid,2-methylsulfanyl pyridine-3-boronic acid,2-methylsulfanyl pyridin-3-ylboronic acid,2-methylthio-3-pyridinyl boronic acid,acmc-2098of,boronic acid, b-2-methylthio-3-pyridinyl PubChem CID: 44755183 IUPAC Name: (2-methylsulfanylpyridin-3-yl)boronic acid SMILES: B(C1=C(N=CC=C1)SC)(O)O