accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1073338-92-7

BOOH3CCH3H3CH3COOOCH3

CAS RN 1073338-92-7

IUPAC Name: ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylate
Synonyms: ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylate
Molecular Weight (g/mol): 266.1
Molecular Formula: C13H19BO5
InChi Key: UMMKKZJPSPIDGN-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(O1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results