accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1073354-27-4

BOOH3CCH3H3CH3CNHN

CAS RN 1073354-27-4

IUPAC Name: N-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Synonyms: N-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Weight (g/mol): 310.2
Molecular Formula: C18H23BN2O2
InChi Key: BWIHFPRFKBKWMU-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(NCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results