accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3COCH3

CAS RN 1073371-72-8

IUPAC Name: 2-[3-(4-methoxyphenyl)propyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[3-(4-methoxyphenyl)propyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 276.18
Molecular Formula: C16H25BO3
InChi Key: ZJNJYHQBUIHZLN-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCCB2OC(C)(C)C(C)(C)O2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results