accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH2O

CAS RN 107377-07-1

IUPAC Name: 4-butylbenzamide
Synonyms: 4-butylbenzamide 丁苯柳胺
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: KFINRIKJBULSTD-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results