accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHBr

CAS RN 1074-16-4

IUPAC Name: 2-(2-bromophenyl)ethan-1-ol
Synonyms: 2-(2-bromophenyl)ethanol
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: ADLOWZRDUHSVRU-UHFFFAOYSA-N
SMILES: OCCC1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results