accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3BrCH3Br

CAS RN 1074-24-4

IUPAC Name: 1,4-dibromo-2,5-dimethylbenzene
Synonyms: 1,4-dibromo-2,5-dimethylbenzene
Molecular Weight (g/mol): 263.96
Molecular Formula: C8H8Br2
InChi Key: QENIALCDPFDFHX-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(C)C=C1Br
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)