CAS RN 1074-61-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(4-Vinylphenyl)methanol (~0.1% TBC stabilizer), TRC
CAS: 1074-61-9 Molecular Formula: C9 H10 O Molecular Weight (g/mol): 134.18 Synonym: Benzyl alcohol, p-vinyl- (6CI,7CI,8CI),4-Ethenylbenzenemethanol,(4-Ethenylphenyl)methanol,4-(Hydroxymethyl)styrene,4-Vinylbenzenemethanol,4-Vinylbenzyl alcohol,p-(Hydroxymethyl)styrene,p-Vinylbenzyl alcohol IUPAC Name: (4-vinylphenyl)methanol SMILES: OCc1ccc(C=C)cc1